Tisk…

Potassium (cyclopropylmethyl)trifluoroborate 95%

Dodavatel: Apollo Scientific

Synonyms: potassium cyclopropylmethyl(trifluoro)boranuide

PC905207-1G PC905207-250MG
APOSPC905207-1GEA 7110 CZK
APOSPC905207-1G APOSPC905207-250MG
Potassium (cyclopropylmethyl)trifluoroborate 95%
Potassium (cyclopropylmethyl)trifluoroborate
Vzorec: C₄H₇BF₃·K
MW: 162,003 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11052655
CAS číslo: 1356481-57-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR