Tisk…

Potassium tert-butyl N-[2-(trifluoroboranuidyl)ethyl]carbamate 95%

Dodavatel: Apollo Scientific

Synonyms: Potassium, Potassium 2-(Boc-aminoethyl)trifluoroborate, Potassium (2-((tert-butoxycarbonyl)amino)ethyl)trifluoroborate, trifluoro-[2-[(2-methylpropan-2-yl)oxycarbonylamino]ethyl]boranuide

PC905181-1G PC905181-5G
APOSPC905181-1GEA 2690 CZK
APOSPC905181-1G APOSPC905181-5G
Potassium tert-butyl N-[2-(trifluoroboranuidyl)ethyl]carbamate 95%
Potassium tert-butyl N-[2-(trifluoroboranuidyl)ethyl]carbamate
Vzorec: C₇H₁₄BF₃NO₂·K
MW: 251,096 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10698613
CAS číslo: 926280-83-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR