2-(4-Fluoro-1H-indol-1-yl)acetic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC905151-1G
PC905151-250MG
APOSPC905151-1GEA
4300
CZK
APOSPC905151-1G
APOSPC905151-250MG
2-(4-Fluoro-1H-indol-1-yl)acetic acid 97%
2-(4-Fluoro-1H-indol-1-yl)acetic acid
Vzorec:
C₁₀H₈FNO₂ MW: 193,174 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19441897 CAS číslo: 1313712-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...