Tisk…

4-Fluoro-1H-indole-3-carbaldehyde 95%

Dodavatel: Apollo Scientific

PC905140-1G PC905140-250MG
APOSPC905140-1GEA 3110 CZK
APOSPC905140-1G APOSPC905140-250MG
4-Fluoro-1H-indole-3-carbaldehyde 95%
4-Fluoro-1H-indole-3-carbaldehyde
Vzorec: C₉H₆FNO
MW: 163,148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00056930
CAS číslo: 23073-31-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR