Tisk…

7-Fluoro-benzo[d]isoxazol-3-ylamine 98%

Dodavatel: Apollo Scientific

PC905115-1G PC905115-250MG
APOSPC905115-1GEA 12010 CZK
APOSPC905115-1G APOSPC905115-250MG
7-Fluoro-benzo[d]isoxazol-3-ylamine 98%
7-Fluoro-benzo[d]isoxazol-3-ylamine
Vzorec: C₇H₅FN₂O
MW: 152,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845366
CAS číslo: 268734-39-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR