1-(5-Fluoro-2-nitrophenyl)ethanone 95%
Dodavatel: Apollo Scientific
PC905105-5G
APOSPC905105-5GEA
3540
CZK
APOSPC905105-5G
1-(5-Fluoro-2-nitrophenyl)ethanone 95%
1-(5-Fluoro-2-nitrophenyl)ethanone
Vzorec:
C₈H₆FNO₃ MW: 183,137 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835330 CAS číslo: 2250-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...