Tisk…

5-Fluoro-4-chromanone 95%

Dodavatel: Apollo Scientific

PC905030-1G PC905030-5G PC905030-100MG PC905030-250MG
APOSPC905030-1GEA 9690 CZK
APOSPC905030-1G APOSPC905030-5G APOSPC905030-100MG APOSPC905030-250MG
5-Fluoro-4-chromanone 95%
5-Fluoro-4-chromanone
Vzorec: C₉H₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD09743992
CAS číslo: 188826-32-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR