2-(6-Fluoro-pyridin-2-ylamino)-ethanol 97%
Dodavatel: Apollo Scientific
PC905021-1G
PC905021-5G
APOSPC905021-1GEA
6020
CZK
APOSPC905021-1G
APOSPC905021-5G
2-(6-Fluoro-pyridin-2-ylamino)-ethanol 97%
2-(6-Fluoro-pyridin-2-ylamino)-ethanol
Vzorec:
C₇H₉FN₂O MW: 156.158 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD14600159 CAS číslo: 1000981-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...