4-Fluoro-2-morpholinoaniline 95%
Dodavatel: Apollo Scientific
PC904965-1G
PC904965-5G
APOSPC904965-1GEA
2570
CZK
APOSPC904965-1G
APOSPC904965-5G
4-Fluoro-2-morpholinoaniline 95%
4-Fluoro-2-morpholinoaniline
Vzorec:
C₁₀H₁₃FN₂O MW: 196,221 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11849334 CAS číslo: 232951-87-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...