Tisk…

5-Fluoro-2,N-dimethylaniline 98%

Dodavatel: Apollo Scientific

PC904944-1G PC904944-5G
APOSPC904944-1GEA 4140 CZK
APOSPC904944-1G APOSPC904944-5G
5-Fluoro-2,N-dimethylaniline 98%
5-Fluoro-2,N-dimethylaniline
Vzorec: C₈H₁₀FN
MW: 139,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10000410
CAS číslo: 881848-77-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR