Tisk…

2-(2-Fluorophenyl)cyclopropanecarboxylic acid 95%

Dodavatel: Apollo Scientific

PC904926-1G PC904926-250MG
APOSPC904926-1GEA 7910 CZK
APOSPC904926-1G APOSPC904926-250MG
2-(2-Fluorophenyl)cyclopropanecarboxylic acid 95%
2-(2-Fluorophenyl)cyclopropanecarboxylic acid
Vzorec: C₁₀H₉FO₂
MW: 180,176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11895893
CAS číslo: 455267-56-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR