Tisk…

Methyl 2-amino-4-bromo-5-fluorobenzoate 95+%

Dodavatel: Apollo Scientific
Varování

PC904903-1G PC904903-250MG PC904903-5G
APOSPC904903-1GEA 1540 CZK
APOSPC904903-1G APOSPC904903-250MG APOSPC904903-5G
Methyl 2-amino-4-bromo-5-fluorobenzoate 95+%
Methyl 2-amino-4-bromo-5-fluorobenzoate
Vzorec: C₈H₇BrFNO₂
MW: 248,049 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22565660
CAS číslo: 1395493-30-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR