(4-Fluoro-3-nitrophenyl)methanamine 95%
Dodavatel: Apollo Scientific
PC904897-1G
PC904897-250MG
APOSPC904897-1GEA
24600
CZK
APOSPC904897-1G
APOSPC904897-250MG
(4-Fluoro-3-nitrophenyl)methanamine 95%
(4-Fluoro-3-nitrophenyl)methanamine
Vzorec:
C₇H₇FN₂O₂ MW: 170,141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06212995 CAS číslo: 771581-73-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...