N-(3-Fluoro-2-chloro-phenyl)-formamide 95%
Dodavatel: Apollo Scientific
PC904889-1G
PC904889-250MG
APOSPC904889-1GEA
12400
CZK
APOSPC904889-1G
APOSPC904889-250MG
N-(3-Fluoro-2-chloro-phenyl)-formamide 95%
N-(3-Fluoro-2-chloro-phenyl)-formamide
Vzorec:
C₇H₅ClFNO MW: 173,572 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19441850 CAS číslo: 1070892-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...