Tisk…

Methyl 2-fluoro-3-hydroxybenzoate 95%

Dodavatel: Apollo Scientific

PC904877-1G
APOSPC904877-1GEA 3020 CZK
APOSPC904877-1G
Methyl 2-fluoro-3-hydroxybenzoate 95%
Methyl 2-fluoro-3-hydroxybenzoate
Vzorec: C₈H₇FO₃
MW: 170,138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698042
CAS číslo: 1214324-64-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR