Tisk…

5-Fluoro-2-methyl-3-nitroaniline 95%

Dodavatel: Apollo Scientific

PC904806-1G PC904806-250MG
APOSPC904806-1GEA 22950 CZK
APOSPC904806-1G APOSPC904806-250MG
5-Fluoro-2-methyl-3-nitroaniline 95%
5-Fluoro-2-methyl-3-nitroaniline
Vzorec: C₇H₇FN₂O₂
MW: 170,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08690074
CAS číslo: 168770-44-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR