Tisk…

(R)-N-(2,2-Difluoroethylidene)-1-phenylethylamine 95%

Dodavatel: Apollo Scientific

PC904768-1G PC904768-250MG
APOSPC904768-1GEA 13860 CZK
APOSPC904768-1G APOSPC904768-250MG
(R)-N-(2,2-Difluoroethylidene)-1-phenylethylamine 95%
(R)-N-(2,2-Difluoroethylidene)-1-phenylethylamine
Vzorec: C₁₀H₁₁F₂N
MW: 183,198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01321162
CAS číslo: 160797-29-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR