Tisk…

(3,5-Difluoro-4-nitrophenyl)methanol 95%

Dodavatel: Apollo Scientific

PC904722-1G PC904722-250MG
APOSPC904722-1GEA 2530 CZK
APOSPC904722-1G APOSPC904722-250MG
(3,5-Difluoro-4-nitrophenyl)methanol 95%
(3,5-Difluoro-4-nitrophenyl)methanol
Vzorec: C₇H₅F₂NO₃
MW: 189,116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18633079
CAS číslo: 1123172-89-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR