Tisk…

(S)-6-Fluorochroman-4-amine hydrochloride 97+%

Dodavatel: Apollo Scientific
Varování

Synonyms: (4S)-6-Fluoro-3,4-dihydro-2H-chromen-4-amine hydrochloride

PC904702-1G PC904702-250MG
APOSPC904702-1GEA 9330 CZK
APOSPC904702-1G APOSPC904702-250MG
(S)-6-Fluorochroman-4-amine hydrochloride 97+%
(S)-6-Fluorochroman-4-amine hydrochloride
Vzorec: C₉H₁₀FNO·ClH
MW: 203,641 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15474861
CAS číslo: 1260609-97-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR