Tisk…

4-(Trifluoromethyl)benzyl isocyanate 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Trifluoromethylbenzylisocyanate, 1-(Isocyanatomethyl)-4-(trifluoromethyl)benzene

PC904664-5G
APOSPC904664-5GEA 3970 CZK
APOSPC904664-5G
4-(Trifluoromethyl)benzyl isocyanate 95%
4-(Trifluoromethyl)benzyl isocyanate

May form small amount of sediment

Vzorec: C₉H₆F₃NO
MW: 201.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07779315
CAS číslo: 102422-55-9
UN: 2206
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR