Tisk…

Pyridine trifluoroacetate 98%

Dodavatel: Apollo Scientific

Synonyms: Pyridine 2,2,2-trifluoroacetate

PC904612-100G PC904612-25G PC904612-500G
APOSPC904612-100GEA 2360 CZK
APOSPC904612-100G APOSPC904612-25G APOSPC904612-500G
Pyridine trifluoroacetate 98%
Pyridine trifluoroacetate
Vzorec: CF₃CO₂H·C₅H₅N
MW: 193,123 g/mol
Bod tání: 83…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00013072
CAS číslo: 464-05-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR