Tisk…

6-Bromo-5-fluoro-3-methyl-1H-indazole 96%

Dodavatel: Apollo Scientific
Varování

PC904604-100MG PC904604-250MG
APOSPC904604-100MGEA 1620 CZK
APOSPC904604-100MG APOSPC904604-250MG
6-Bromo-5-fluoro-3-methyl-1H-indazole 96%
6-Bromo-5-fluoro-3-methyl-1H-indazole
Vzorec: C₈H₆BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD20278282
CAS číslo: 1394120-64-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR