Tisk…

2-(Trifluoromethyl)-1H-imidazole 95%

Dodavatel: Apollo Scientific

PC904601-1G
APOSPC904601-1GEA 2360 CZK
APOSPC904601-1G
2-(Trifluoromethyl)-1H-imidazole 95%
2-(Trifluoromethyl)-1H-imidazole
Vzorec: C₄H₃F₃N₂
MW: 136,075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18415320
CAS číslo: 66675-22-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR