6-Fluoro-3-methyl-1H-pyrazolo[3,4-b]pyridine 95%
Dodavatel: Apollo Scientific
PC904597-1G
PC904597-250MG
APOSPC904597-1GEA
22950
CZK
APOSPC904597-1G
APOSPC904597-250MG
6-Fluoro-3-methyl-1H-pyrazolo[3,4-b]pyridine 95%
6-Fluoro-3-methyl-1H-pyrazolo[3,4-b]pyridine
Vzorec:
C₇H₆FN₃ MW: 151,141 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11616901 CAS číslo: 920036-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...