Tisk…

2-Fluoro-3-(methoxycarbonyl)benzoic acid 98%

Dodavatel: Apollo Scientific

PC904567-1G PC904567-250MG
APOSPC904567-1GEA 17820 CZK
APOSPC904567-1G APOSPC904567-250MG
2-Fluoro-3-(methoxycarbonyl)benzoic acid 98%
2-Fluoro-3-(methoxycarbonyl)benzoic acid
Vzorec: C₉H₇FO₄
Storage Temperature: Ambient
MDL Number: MFCD12406363
CAS číslo: 914301-44-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR