(S)-2-(Hydroxymethyl)-4,4-difluoropyrrolidine hydrochloride 95%
Dodavatel: Apollo Scientific
Synonyms:
(S)-(4,4-Difluoropyrrolidin-2-yl)methanol hydrochloride
PC904562-1G
PC904562-250MG
APOSPC904562-1GEA
7540
CZK
APOSPC904562-1G
APOSPC904562-250MG
(S)-2-(Hydroxymethyl)-4,4-difluoropyrrolidine hydrochloride 95%
(S)-2-(Hydroxymethyl)-4,4-difluoropyrrolidine hydrochloride
Vzorec:
C₅H₉F₂NO·ClH MW: 173,589 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22570251 CAS číslo: 623583-10-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...