(5-(Trifluoromethyl)pyridin-2-yl)methanamine 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC904531-1G
PC904531-500MG
PC904531-5G
APOSPC904531-1GEA
11810
CZK
APOSPC904531-1G
APOSPC904531-500MG
APOSPC904531-5G
(5-(Trifluoromethyl)pyridin-2-yl)methanamine 95+%
(5-(Trifluoromethyl)pyridin-2-yl)methanamine
Vzorec:
C₇H₇F₃N₂ MW: 176,139 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04117761 CAS číslo: 164341-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...