Tisk…

2-Fluoro-N-methyl-5-nitroaniline 95%

Dodavatel: Apollo Scientific

PC904455-10G PC904455-1G PC904455-5G
APOSPC904455-10GEA 8040 CZK
APOSPC904455-10G APOSPC904455-1G APOSPC904455-5G
2-Fluoro-N-methyl-5-nitroaniline 95%
2-Fluoro-N-methyl-5-nitroaniline
Vzorec: C₇H₇FN₂O₂
MW: 170,141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527248
CAS číslo: 110729-51-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR