Tisk…

tert-Butyl n-(3,3-difluorocyclobutyl)carbamate

Dodavatel: Apollo Scientific

PC904377-25G PC904377-5G
APOSPC904377-25GEA 13060 CZK
APOSPC904377-25G APOSPC904377-5G
tert-Butyl n-(3,3-difluorocyclobutyl)carbamate
tert-Butyl n-(3,3-difluorocyclobutyl)carbamate
Vzorec: C₉H₁₅F₂NO₂
MW: 207.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10001483
CAS číslo: 1029720-19-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR