Tisk…

Methyl 3-hydroxy-3-(trifluoromethyl)cyclobutane-1-carboxylate

Dodavatel: Apollo Scientific

PC904340-10G PC904340-1G PC904340-5G
APOSPC904340-10GEA 40670 CZK
APOSPC904340-10G APOSPC904340-1G APOSPC904340-5G
Methyl 3-hydroxy-3-(trifluoromethyl)cyclobutane-1-carboxylate
Methyl 3-hydroxy-3-(trifluoromethyl)cyclobutane-1-carboxylate
Vzorec: C₇H₉F₃O₃
MW: 198,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23106188
CAS číslo: 1392803-31-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR