4-(Chloromethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole 95%
Dodavatel: Apollo Scientific
PC904276-1G
PC904276-500MG
APOSPC904276-1GEA
20470
CZK
APOSPC904276-1G
APOSPC904276-500MG
4-(Chloromethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole 95%
4-(Chloromethyl)-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Vzorec:
C₆H₆ClF₃N₂ MW: 198,573 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21647876 CAS číslo: 1222556-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...