Tisk…

N-Isopropyl-3-fluoroaniline 95%

Dodavatel: Apollo Scientific

PC904100-1G PC904100-250MG
APOSPC904100-1GEA 15310 CZK
APOSPC904100-1G APOSPC904100-250MG
N-Isopropyl-3-fluoroaniline 95%
N-Isopropyl-3-fluoroaniline
Vzorec: C₉H₁₂FN
MW: 153,197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972886
CAS číslo: 121431-27-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR