Tisk…
Ethyl 2,2-difluoro-3-aminopropanoate hydrochloride 95%
Dodavatel: Apollo Scientific
Synonyms: Ethyl 3-amino-2,2-difluoropropanoate hydrochloride
PC904025-1G PC904025-250MG
APOSPC904025-1GEA 11080 CZK
APOSPC904025-1G APOSPC904025-250MG
Ethyl 2,2-difluoro-3-aminopropanoate hydrochloride 95%
Ethyl 2,2-difluoro-3-aminopropanoate hydrochloride
Vzorec: C₅H₉F₂NO₂·ClH
MW: 189.588 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185039
CAS číslo: 1263284-04-3