Tisk…

8-Fluoroquinoline-2-carboxylic acid 95%

Dodavatel: Apollo Scientific

PC904009-1G PC904009-250MG
APOSPC904009-1GEA 9720 CZK
APOSPC904009-1G APOSPC904009-250MG
8-Fluoroquinoline-2-carboxylic acid 95%
8-Fluoroquinoline-2-carboxylic acid
Vzorec: C₁₀H₆FNO₂
MW: 191.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08690746
CAS číslo: 914208-13-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR