Tisk…

1-Acetyl-5-methyl-3-(trifluoromethyl)pyrazole

Dodavatel: Apollo Scientific
Varování

PC9040-10G PC9040-5G
APOSPC9040-10GEA 7180 CZK
APOSPC9040-10G APOSPC9040-5G
1-Acetyl-5-methyl-3-(trifluoromethyl)pyrazole
1-Acetyl-5-methyl-3-(trifluoromethyl)pyrazole
Vzorec: C₇H₇F₃N₂O
MW: 192,138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153637
CAS číslo: 959573-03-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR