5-Methoxy-2-(trifluoromethyl)benzoic acid 98%
Dodavatel: Apollo Scientific
PC903941-10G
PC903941-1G
PC903941-5G
APOSPC903941-10GEA
29790
CZK
APOSPC903941-10G
APOSPC903941-1G
APOSPC903941-5G
5-Methoxy-2-(trifluoromethyl)benzoic acid 98%
5-Methoxy-2-(trifluoromethyl)benzoic acid
Vzorec:
C₉H₇F₃O₃ MW: 220.145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185683 CAS číslo: 716-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...