Tisk…

2-Fluoro-4-iodo-1-methoxybenzene 95%

Dodavatel: Apollo Scientific

PC903918-10G PC903918-1G PC903918-5G
APOSPC903918-10GEA 29330 CZK
APOSPC903918-10G APOSPC903918-1G APOSPC903918-5G
2-Fluoro-4-iodo-1-methoxybenzene 95%
2-Fluoro-4-iodo-1-methoxybenzene
Vzorec: C₇H₆FIO
MW: 252.025 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00691526
CAS číslo: 3824-21-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR