Tisk…

1-Methyl-5-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde

Dodavatel: Apollo Scientific
Varování

PC903913-1G PC903913-5G
APOSPC903913-1GEA 3740 CZK
APOSPC903913-1G APOSPC903913-5G
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
1-Methyl-5-(trifluoromethyl)-1H-pyrazole-4-carbaldehyde
Vzorec: C₆H₅F₃N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11707230
CAS číslo: 497833-04-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR