Tisk…

4-(2-Fluoro-phenyl)-piperidin-4-ol 95%

Dodavatel: Apollo Scientific

PC903851-1G PC903851-250MG
APOSPC903851-1GEA 16340 CZK
APOSPC903851-1G APOSPC903851-250MG
4-(2-Fluoro-phenyl)-piperidin-4-ol 95%
4-(2-Fluoro-phenyl)-piperidin-4-ol
Vzorec: C₁₁H₁₄FNO
MW: 195,233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05237198
CAS číslo: 871113-19-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR