5-Chloro-2-(chloromethyl)-3-fluoropyridine 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC903848-1G
PC903848-250MG
APOSPC903848-1GEA
15110
CZK
APOSPC903848-1G
APOSPC903848-250MG
5-Chloro-2-(chloromethyl)-3-fluoropyridine 97%
5-Chloro-2-(chloromethyl)-3-fluoropyridine
Vzorec:
C₆H₄Cl₂FN MW: 180.007 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16608217 CAS číslo: 1227585-36-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...