Tisk…

Potassium cyclopentylmethyltrifluoroborate 96%

Dodavatel: Apollo Scientific

Synonyms: Potassium (cyclopentylmethyl)trifluoroborate

PC903841-1G PC903841-5G
APOSPC903841-1GEA 3110 CZK
APOSPC903841-1G APOSPC903841-5G
Potassium cyclopentylmethyltrifluoroborate 96%
Potassium cyclopentylmethyltrifluoroborate
Vzorec: C₆H₁₁BF₃·K
Storage Temperature: Freezer
MDL Number: MFCD11052656
CAS číslo: 331282-37-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR