Tisk…

2-(2-Fluoropyridin-3-yl)propan-2-ol 95%

Dodavatel: Apollo Scientific

PC903803-1G PC903803-5G
APOSPC903803-1GEA 4370 CZK
APOSPC903803-1G APOSPC903803-5G
2-(2-Fluoropyridin-3-yl)propan-2-ol 95%
2-(2-Fluoropyridin-3-yl)propan-2-ol
Vzorec: C₈H₁₀FNO
MW: 155,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11977651
CAS číslo: 40247-48-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR