Tisk…

8-Bromo-5-fluoroquinolin-4(1H)-one

Dodavatel: Apollo Scientific
Varování

PC903774-1G PC903774-250MG
APOSPC903774-1GEA 7250 CZK
APOSPC903774-1G APOSPC903774-250MG
8-Bromo-5-fluoroquinolin-4(1H)-one
8-Bromo-5-fluoroquinolin-4(1H)-one
Vzorec: C₉H₅BrFNO
MW: 242,045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527288
CAS číslo: 1065092-35-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR