Tisk…

1-(5-Fluoropyridin-3-yl)ethanone 96%

Dodavatel: Apollo Scientific
Varování

PC903756-100MG PC903756-250MG
APOSPC903756-100MGEA 827 CZK
APOSPC903756-100MG APOSPC903756-250MG
1-(5-Fluoropyridin-3-yl)ethanone 96%
1-(5-Fluoropyridin-3-yl)ethanone
Vzorec: C₇H₆FNO
MW: 139,127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375075
CAS číslo: 342602-55-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR