Tisk…

3,5-Difluoro-4-methylbenzonitrile 95%

Dodavatel: Apollo Scientific

PC903678-1G PC903678-5G
APOSPC903678-1GEA 3940 CZK
APOSPC903678-1G APOSPC903678-5G
3,5-Difluoro-4-methylbenzonitrile 95%
3,5-Difluoro-4-methylbenzonitrile
Vzorec: C₈H₅F₂N
MW: 153,129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835197
CAS číslo: 170572-50-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR