Tisk…

Methyl 4-fluoro-1H-pyrrole-2-carboxylate 98%

Dodavatel: Apollo Scientific

PC903659-1G PC903659-250MG
APOSPC903659-1GEA 2730 CZK
APOSPC903659-1G APOSPC903659-250MG
Methyl 4-fluoro-1H-pyrrole-2-carboxylate 98%
Methyl 4-fluoro-1H-pyrrole-2-carboxylate
Vzorec: C₆H₆FNO₂
MW: 143,116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042594
CAS číslo: 475561-89-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR