Tisk…

1-Fluoro-3-phenoxybenzene 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Fluorodiphenyl ether

PC903022-1G PC903022-5G
APOSPC903022-1GEA 6220 CZK
APOSPC903022-1G APOSPC903022-5G
1-Fluoro-3-phenoxybenzene 95%
1-Fluoro-3-phenoxybenzene
Vzorec: C₁₂H₉FO
MW: 188,198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015805
CAS číslo: 3798-89-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR