2-Chloro-4-fluoro-3-nitrotoluene 98%
Dodavatel: Apollo Scientific
PC902965-1G
PC902965-250MG
APOSPC902965-1GEA
18420
CZK
APOSPC902965-1G
APOSPC902965-250MG
2-Chloro-4-fluoro-3-nitrotoluene 98%
2-Chloro-4-fluoro-3-nitrotoluene
Vzorec:
C₇H₅ClFNO₂ MW: 189,572 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26940372 CAS číslo: 1805115-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...