2-Fluoro-4-(1-methylhydrazin-1-yl)benzonitrile 96%
Dodavatel: Apollo Scientific
PC902823-1G
PC902823-5G
APOSPC902823-1GEA
6220
CZK
APOSPC902823-1G
APOSPC902823-5G
2-Fluoro-4-(1-methylhydrazin-1-yl)benzonitrile 96%
2-Fluoro-4-(1-methylhydrazin-1-yl)benzonitrile
Vzorec:
C₈H₈FN₃ MW: 165,168 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22549309 CAS číslo: 1357626-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...