Tisk…

5-Chloro-2-fluoro-3-hydroxypyridine 95%

Dodavatel: Apollo Scientific

PC902807-1G PC902807-250MG
APOSPC902807-1GEA 14290 CZK
APOSPC902807-1G APOSPC902807-250MG
5-Chloro-2-fluoro-3-hydroxypyridine 95%
5-Chloro-2-fluoro-3-hydroxypyridine
Vzorec: C₅H₃ClFNO
MW: 147,535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835255
CAS číslo: 1003711-65-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR